Merck
  • Plasma Membrane Na⁺-Coupled Citrate Transporter (SLC13A5) and Neonatal Epileptic Encephalopathy.
Plasma Membrane Na⁺-Coupled Citrate Transporter (SLC13A5) and Neonatal Epileptic Encephalopathy.
Molecules (Basel, Switzerland) (2017-03-08)
Yangzom D Bhutia, Jonathan J Kopel, John J Lawrence, Volker Neugebauer, Vadivel Ganapathy
ABSTRACT
SLC13A5 is a Na⁺-coupled transporter for citrate that is expressed in the plasma membrane of specific cell types in the liver, testis, and brain. It is an electrogenic transporter with a Na⁺:citrate