Merck
  • (-)-Epigallocatechin gallate but not chlorogenic acid upregulates osteoprotegerin synthesis through regulation of bone morphogenetic protein-4 in osteoblasts.
(-)-Epigallocatechin gallate but not chlorogenic acid upregulates osteoprotegerin synthesis through regulation of bone morphogenetic protein-4 in osteoblasts.
Experimental and therapeutic medicine (2017-07-05)
Kazuhiko Fujita, Takanobu Otsuka, Naohiro Yamamoto, Shingo Kainuma, Reou Ohguchi, Tetsu Kawabata, Go Sakai, Gen Kuroyanagi, Rie Matsushima-Nishiwaki, Osamu Kozawa, Haruhiko Tokuda
ABSTRACT
Chlorogenic acid (CGA) is a primary phenolic component of coffee and (-)-epigallocatechin gallate (EGCG) is a primary flavonoid component of green tea, both of which have been documented to possess beneficial health properties. A previous study by the present authors demonstrated that p38 mitogen-activated protein kinase (MAPK) may be associated with osteoprotegerin synthesis stimulated by bone morphogenetic protein-4 (BMP-4) in osteoblast-like MC3T3-E1 cells. In the present study, the effects of CGA and EGCG on BMP-4-stimulated osteoprotegerin synthesis in MC3T3-E1 cells were investigated. It was observed that CGA had no effect on osteoprotegerin release stimulated by BMP-4, whereas EGCG significantly enhanced BMP-4-stimulated osteoprotegerin release (P=0.003). Levels of osteoprotegerin mRNA expression induced by BMP-4 were also significantly increased by EGCG (P=0.03). By contrast, EGCG had no significant effect on phosphorylation of Smad1 or p38 MAPK induced by BMP-4. In addition, EGCG had little effect on BMP-induced phosphorylation of p70 S6 kinase; however rapamycin, as an inhibitor of p70 S6 kinase, significantly suppressed osteoprotegerin release (P=0.007). These data suggest that EGCG but not CGA may upregulate the synthesis of osteoprotegerin induced by BMP-4 in osteoblasts.