Merck
  • Smartphone-based enzymatic biosensor for oral fluid L-lactate detection in one minute using confined multilayer paper reflectometry.
Smartphone-based enzymatic biosensor for oral fluid L-lactate detection in one minute using confined multilayer paper reflectometry.
Biosensors & bioelectronics (2017-03-08)
Donato Calabria, Cristiana Caliceti, Martina Zangheri, Mara Mirasoli, Patrizia Simoni, Aldo Roda
ABSTRACT
The development of smartphone-based biosensors for point-of-care testing (POCT) applications allows realizing "all in one" instruments, with large potential distribution among the general population. With this respect, paper color-based detection performed by reflectance measurement is the most popular, simple, inexpensive and straightforward method. Despite the large number of scientific publications related to these biosensors, they still suffer from a poor detectability and reproducibility related to inhomogeneity of color development, which leads to low assay reproducibility. To overcome these problems, we propose a smartphone paper-based biosensor, in which all the reagents necessary to complete the analysis are co-entrapped on paper in a "wafer"-like bilayer film of polyelectrolytes (Poly (allyl amine hydrochloride/poly(sodium 4-styrene sulfonate)). Using a 3D printing low-cost technology we fabricated the smartphone-based device that consists in a cover accessory attached to the smartphone and incorporating a light diffuser over the flash to improve the image quality, a mini dark box and a disposable analytical cartridge containing all the reagents necessary for the complete analysis. The biosensor was developed exploiting coupled enzyme reactions for quantifying L-lactate in oral fluid, which is considered a biomarker of poor tissue perfusion, a key element in the management of severe sepsis, septic shock and in sports performance evaluation. The developed method is sensitive, rapid, and it allows detecting L-lactate in oral fluid in the relevant physiological range, with a limit of detection of 0.1mmolL