Merck
  • Perturbation of Akt Signaling, Mitochondrial Potential, and ADP/ATP Ratio in Acidosis-Challenged Rat Cortical Astrocytes.
Perturbation of Akt Signaling, Mitochondrial Potential, and ADP/ATP Ratio in Acidosis-Challenged Rat Cortical Astrocytes.
Journal of cellular biochemistry (2016-09-09)
King-Chuen Wu, Ka-Shun Cheng, Yu-Wen Wang, Yuh-Fung Chen, Kar-Lok Wong, Tzu-Hui Su, Paul Chan, Yuk-Man Leung
ABSTRACT
Cells switch to anaerobic glycolysis when there is a lack of oxygen during brain ischemia. Extracellular pH thus drops and such acidosis causes neuronal cell death. The fate of astrocytes, mechanical, and functional partners of neurons, in acidosis is less studied. In this report, we investigated the signaling in acidosis-challenged rat cortical astrocytes and whether these signals were related to mitochondrial dysfunction and cell death. Exposure to acidic pH (6.8, 6.0) caused Ca