Merck
Synthesis and stability studies of Ga-67 labeled phosphonium salts.
Journal of labelled compounds & radiopharmaceuticals (2017-01-13)
Mingyue Kardashinsky, Nigel Lengkeek, Louis M Rendina
ABSTRACT
Delocalized lipophilic cations such as tri- and tetra-arylphosphonium are able to diffuse across the mitochondrial membrane, which allows them to selectively accumulate in cells with a high transmembrane potential (ΔΨ