Merck
Whole-mount immunoelectron tomography of chromosomes and cells.
Methods in molecular biology (Clifton, N.J.) (2007-07-28)
Peter Engelhardt, Jari Meriläinen, Fang Zhao, Susumu Uchiyama, Kiichi Fukui, Veli-Pekka Lehto
ABSTRACT
Standard immunogold-labeling methods in transmission electron microscopy (TEM) are unable to locate immunogold particles in the depth direction. This inability does not only concern bulky whole mounts, but also sections. A partial solution to the problem is stereo inspection. However, three-dimensional reconstruction of immunogold-labeled structures, that is, immuno-electron tomography (IET), is a correct solution for this inconsistency. Striking improvement in resolution is achieved: the 1.4-nm immunogold particles are shown in IET that are not detected in the original tilt series. IET is not restricted to laboratories with advanced medium- or high-voltage TEM and super-computing facilities; the methods we have developed for whole-mounted chromosomes and also for whole-mounted cytoskeleton of fibroblasts work remarkably well with ordinary 80-kV TEMs equipped with a goniometer to collect tilt series for IET on film. In addition, free programs are available to produce three-dimensional reconstructions even without high-performance computers. These improvements make it possible to many laboratories without modern facilities to perform IET reconstruction with standard TEM apparatus.