Merck
Fetuin A stabilizes m-calpain and facilitates plasma membrane repair.
The Journal of biological chemistry (2007-10-19)
Ronald L Mellgren, Xinhua Huang
ABSTRACT
Yeast two-hybrid experiments identified alpha(2)-Heremans-Schmid glycoprotein (human fetuin A) as a binding partner for calpain domain III (DIII). The tandem DIIIs of calpain-10 interacted under the most selective culture conditions, but DIIIs of m-calpain, calpain-3, and calpain-5 also interacted under less stringent selection. DIIIs of mu-calpain, calpain-6, and the tandem DIII-like domains of the Dictyostelium Cpl protein did not interact with alpha(2)-Heremans-Schmid glycoprotein in the yeast two-hybrid system. Bovine fetuin A stabilized proteolytic activity of purified m-calpain incubated in the presence of mm calcium chloride and prevented calcium-dependent m-calpain aggregation. Consistent with the yeast two-hybrid studies, fetuin A neither stabilized mu-calpain nor prevented its aggregation. Confocal immunofluorescence microscopy of scratch-damaged L6 myotubes demonstrated accumulation of m-calpain at the wound site in association with the membrane repair protein, dysferlin. m-Calpain also co-localized with fluorescein-labeled fetuin A at the wound site. The effect of fetuin A on calpain-mediated plasma membrane resealing was investigated using fibroblasts from Capns1(-/-) and Capns1(+/+) mouse embryos. Capns1 encodes the small noncatalytic subunit that is required for the proteolytic function of m- and mu-calpains. Thus, Capns1(-/-) fibroblasts do not express these calpains in active form. Fetuin A increased resealing of scrape-damaged wild-type fibroblasts but not Capns1(-/-) fibroblasts. These studies identify fetuin A as a potential extracellular regulator of m-calpain at nascent sites of plasma membrane wounding.