Merck
  • Stanniocalcin-1 ameliorates lipopolysaccharide-induced pulmonary oxidative stress, inflammation, and apoptosis in mice.
Stanniocalcin-1 ameliorates lipopolysaccharide-induced pulmonary oxidative stress, inflammation, and apoptosis in mice.
Free radical biology & medicine (2014-04-02)
Shih-En Tang, Chin-Pyng Wu, Shu-Yu Wu, Chung-Kan Peng, Wann-Cherng Perng, Bor-Hwang Kang, Shi-Jye Chu, Kun-Lun Huang
ABSTRACT
Stanniocalcin-1 (STC1) is an endogenous glycoprotein whose anti-inflammatory effects occur through induction of uncoupling proteins to reduce oxidative stress. In this study, we tested the hypothesis that exogenous recombinant human STC1 (rhSTC1) protects against lipopolysaccharide (LPS)-induced acute lung injury in mice. Anesthetized C57BL/6 mice underwent intratracheal spraying of LPS (20 µg/10 g body wt), and lung injury was assessed 24h later by analyzing pulmonary edema, bronchoalveolar lavage fluid, and lung histopathology. Lung inflammation, oxidative stress, and expression of STC1 and its downstream uncoupling protein 2 (UCP2) were analyzed at specific time points. Expression of UCP2 was suppressed initially but was subsequently upregulated after STC1 elevation in response to intratracheal administration of LPS. Intratracheal rhSTC1 treatment 1h before or after LPS spraying significantly attenuated pulmonary inflammation, oxidative stress, cell apoptosis, and acute lung injury. Pretreatment with STC1 short interfering RNA 48 h before LPS spraying inhibited the expression of STC1 and UCP2 and significantly increased the extent of lung injury. These findings suggest that STC1 is an endogenous stress protein that may counteract LPS-induced lung injury by inhibiting the inflammatory cascade and inducing antioxidant and antiapoptotic mechanisms. However, the potential clinical application of STC1 and the direct linkage between UCP2 and LPS-induced lung injury remain to be further investigated.