Merck
  • Lipoxin A4 methyl ester ameliorates cognitive deficits induced by chronic cerebral hypoperfusion through activating ERK/Nrf2 signaling pathway in rats.
Lipoxin A4 methyl ester ameliorates cognitive deficits induced by chronic cerebral hypoperfusion through activating ERK/Nrf2 signaling pathway in rats.
Pharmacology, biochemistry, and behavior (2014-06-10)
Wei Jin, Yanqiu Jia, Lining Huang, Tianjun Wang, Hebo Wang, Yanhong Dong, Hezhen Zhang, Mingyue Fan, Peiyuan Lv
ABSTRACT
Lipoxin A4 (LXA4) is known for its powerful anti-inflammatory function. Current studies in vitro suggest that LXA4 possesses novel antioxidant effect. The aim of this study is to examine whether Lipoxin A4 methyl ester (LXA4 ME) has neuroprotective effects against chronic cerebral hypoperfusion, and if so, whether the effects of LXA4 ME are associated with its potential antioxidant property. Adult male Sprague-Dawley rats were subjected to permanent bilateral common carotid artery occlusion (BCCAO) and randomly assigned into four groups: sham (sham-operated) group, vehicle (BCCAO+normal saline) group, LXA4 ME10 (BCCAO+LXA4 ME 10 ng per day) group and LXA4 ME100 (BCCAO+LXA4 ME 100 ng per day) group. LXA4 ME was administered through intracerebroventricular injection for 2 consecutive weeks. LXA4 ME significantly alleviated spatial learning and memory impairments, as assessed by Morris water maze and inhibited the loss of neurons in the CA1 region of the hippocampus. Biochemically, LXA4 ME phosphorylated extracellular signal regulated kinase (ERK) 1/2 and enhanced nuclear factor erythroid 2-related factor 2 (Nrf2) expression and its nuclear translocation, as well as quinone oxidoreductase 1 (NQO1) expression. LXA4 ME reduced lipid peroxidative production in the hippocampus, as measured by immunohistochemical staining for 4-Hydroxynonenal (4-HNE). In addition, LXA4 ME significantly elevated the ratio of Bcl-2/Bax and decreased cleaved caspase-3 expression in the hippocampus. Therefore, these data suggest that LXA4 ME exerts beneficial effects on the cognitive impairment induced by chronic cerebral hypoperfusion through attenuating oxidative injury and reducing neuronal apoptosis in the hippocampus, which is most likely associated with the activation of ERK/Nrf2 signaling pathway.