Merck
Geranylgeranylacetone protects the heart via caveolae and caveolin-3.
Life sciences (2014-03-04)
Yasuo M Tsutsumi, Rie Tsutsumi, Yousuke T Horikawa, Yoko Sakai, Eisuke Hamaguchi, Yoshihiro Ishikawa, Utako Yokoyama, Asuka Kasai, Noriko Kambe, Katsuya Tanaka
ABSTRACT
Geranylgeranylacetone (GGA) is commonly utilized to protect the gastric mucosa in peptic ulcer disease. Recently GGA has been shown to protect the myocardium from ischemia/reperfusion by activating heat shock proteins. However, the exact mechanism as to how GGA activates these protective proteins is unknown. Caveolae and caveolin-3 (Cav-3) have been implicated in ischemia, anesthetic, and opioid induced cardiac protection. Given the lipophilic nature of GGA it is our hypothesis that GGA induced cardiac protection requires caveolae and Cav-3. We used an in vivo mouse model of ischemia-reperfusion injury and performed biochemical assays in excised hearts. GGA treated control mice revealed increased caveolae formation and caveolin-3 in buoyant fractions, mediating heat shock protein 70 activation. Furthermore, control mice treated with GGA were protected against ischemia/reperfusion injury whereas Cav-3 knockout (Cav-3 KO) mice were not. Troponin levels confirmed myocardial damage. Finally, Cav-3 KO mice treated with GGA were not protected against mitochondrial swelling whereas control mice had significant protection. This study showed that caveolae and caveolin-3 are essential in facilitating GGA induced cardiac protection by optimizing spatial and temporal signaling to the mitochondria.