Merck
  • Chemotherapy for initial induction failures in childhood acute lymphoblastic leukemia: a Children's Oncology Group Study (POG 8764).
Chemotherapy for initial induction failures in childhood acute lymphoblastic leukemia: a Children's Oncology Group Study (POG 8764).
Journal of pediatric hematology/oncology (2012-12-06)
Michael J Joyce, Brad H Pollock, Meenakshi Devidas, George R Buchanan, Bruce Camitta
ABSTRACT
Children with acute lymphocytic leukemia who fail to enter remission have a poor prognosis. In a previous study, 9 of 14 children with induction failure entered remission after teniposide (VM26) plus cytosine arabinoside (Ara-C). We attempted to confirm these results. Twenty children received teniposide (200 mg/m/day IV) for 3 days and cytosine arabinoside (100 mg/m/day continuous IV infusion) for 7 days. There were 3 complete and 3 partial responses. Two additional patients achieved a complete response after a second, shorter course of the same agents. Although VM26 plus Ara-C is an active combination for treatment of acute lymphocytic leukemia induction failure, it does not appear as effective as in the initial report. Better treatments for this problem are needed.