Merck
  • Inhibition of dihydropteridine reductase by novel 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine analogs.
Inhibition of dihydropteridine reductase by novel 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine analogs.
Science (New York, N.Y.) (1984-04-27)
C W Abell, R S Shen, W Gessner, A Brossi
ABSTRACT
Hydroxylated derivatives of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP), a nigrostriatal neurotoxin in humans and primates, noncompetitively inhibited dihydropteridine reductase from human liver and rat striatal synaptosomes in vitro at micromolar concentrations. In contrast, MPTP and its chloro- and norderivatives did not inhibit this enzyme at lower than millimolar concentrations. Dihydropteridine reductase converts dihydrobiopterin to tetrahydrobiopterin, the required cofactor for the hydroxylation of aromatic amino acids during the synthesis of dopamine and serotonin.