Merck
  • The effects of a binary mixture of benzo(a)pyrene and 7H-dibenzo(c,g)carbazole on lung tumors and K-ras oncogene mutations in strain A/J mice.
The effects of a binary mixture of benzo(a)pyrene and 7H-dibenzo(c,g)carbazole on lung tumors and K-ras oncogene mutations in strain A/J mice.
Experimental lung research (2001-04-11)
D L Gray, D Warshawsky, W Xue, R Nines, Y Wang, R Yao, G D Stoner
ABSTRACT
Polycyclic aromatic hydrocarbons (PAH) and N-heterocyclic aromatic hydrocarbons (NHA) are environmental pollutants formed during the incomplete combustion of organic materials. Benzo(a)pyrene (BaP) and 7H-dibenzo(c,g)carbazole (DBC) are well-characterized representatives of the PAH and NHA classes of compunds, respectively. Both are demonstrated carcinogens that frequently co-occur in environmental mixtures. This preliminary study was conducted to investigate the effects of a binary mixture of BaP and DBC on lung carcinogenicity in the strain A/J mouse as manifested by tumor development and mutations in the K-ras gene. Male A/J mice were administered the following single intraperitoneal dose (mg/kg) combinations of BaP and DBC dissolved in a 0.2-mL volume of tricaprylin--10 DBC:10 BaP; 2 DBC:10 BaP; 2 DBC:100 BaP; and 10 DBC: 100 BaP, and each of the compounds alone at the same doses. Mice were sacrificed 8 months after carcinogen treatment and lung tumor multiplicity and K-ras mutations determined (high-dose combination). The combination of DBC and BaP produced fewer tumors than the sum of all tumors produced by each compound acting alone. The frequency of tumors with K-ras mutations was also less in a sample of the 10 DBC:100 BaP treatment group than in the same-dose, single compound-treated animals. The dominant mutations produced by BaP and DBC were expressed in tumors from animals treated with the mixture.