Merck
  • Chronic sinusitis in children with respiratory allergy: the role of antimicrobials.
Chronic sinusitis in children with respiratory allergy: the role of antimicrobials.
The Journal of allergy and clinical immunology (1982-04-01)
G S Rachelefsky, R M Katz, S C Siegel
ABSTRACT
We evaluated the role of antimicrobials in the treatment of chronic maxillary sinusitis in children with respiratory allergy. Night and day cough, nasal obstruction, rhinorrhea, postnasal seen. Eighty-four children were treated in a double-blind manner with either amoxicillin, erythromycin, trimethoprim-sulfamethoxazole, or an antihistamine decongestant (carbinoxamine maleate-pseudoephedrine HCl). Radiographic and clinical responses were best with amoxicillin, but trimethoprim-sulfamethoxazole was an adequate alternative. This study demonstrates that allergic children with chronic sinusitis with associated chronic respiratory symptoms are likely to respond clinically and radiologically with antimicrobial treatment.