- Gas chromatographic determination of clopidol in chicken tissues.
Gas chromatographic determination of clopidol in chicken tissues.
Journal - Association of Official Analytical Chemists (1984-09-01)
L G Ekström, J Kuivinen
PMID6501161
ABSTRACT
A method has been developed for the determination of clopidol residues in chicken tissues. After extraction and cleanup, clopidol is esterified in a 2-phase system to clopidol propionate, which is determined by gas chromatography. The 2-phase system includes, in addition to the clopidol dissolved in methanol, aqueous borax solution, hexane, propionic anhydride, and pyridine. Use of these reagents precludes the use of explosive or carcinogenic chemicals in the derivatization step, and the method is therefore suitable for routine laboratory analysis. Levels of 0.5 ppb clopidol in tissue can be determined.