- Mechanical properties and biocompatibility of two polyepoxy matrices: DGEBA-DDM and DGEBA-IPD.
Mechanical properties and biocompatibility of two polyepoxy matrices: DGEBA-DDM and DGEBA-IPD.
Biomaterials (1987-05-01)
R Berruet, E Vinard, A Calle, H L Tighzert, B Chabert, H Magloire, R Eloy
PMID3607148
ABSTRACT
The aim of this paper was to study the biocompatibility and mechanical properties of materials for orthopaedic and odontologic surgical use. The products used were obtained by polycondensation of a diepoxy resin (DGEBA) with two curing agents (DDM or IPD). The materials present a slight swelling in liquid medium and their thermomechanical properties are hardly affected after 12 month implantation. The absence of molecular desorption in isotonic liquid and human serum confirms their hydrolytic stability and thus their inertia. These materials do not give rise to an intolerance reaction by neighbouring tissues during implantation time (1 d to 12 month).