Merck
  • Teratology study in mice subjected to inhalation of diethylhydroxylamine, nitroethane, and diethylamine hydrogen sulfite.
Teratology study in mice subjected to inhalation of diethylhydroxylamine, nitroethane, and diethylamine hydrogen sulfite.
Environmental research (1978-10-01)
R P Beliles, S L Makris, F Ferguson, C Putman, W Sapanski, N Kelly, K Partymiller, J Heicklen
PMID318511
ABSTRACT
Pregnant female mice were exposed from Days 6 to 17 of pregnancy to 8.9 +/- 2.0 ppm of diethylhydroxylamine and 14.3 +/- 2.0 ppm of nitroethane. Exposures were for 8.25 +/- 2.25 hours/day including weekends. The mice were also exposed continuously for 24 hours a day to the vapor of diethylamine hydrogen sulfite. The exposure produced no effect on the dams compared to the control group. There was no evidence of compound-induced terata, variation in sex ratio, embryotoxicity, or inhibition of fetal growth and development.