Merck
  • Phase III trial of adjuvant 5-fluorouracil and adriamycin versus 5-fluorouracil, adriamycin, and polyadenylic-polyuridylic acid (poly A:U) for locally advanced gastric cancer after curative surgery: final results of 15-year follow-up.
Phase III trial of adjuvant 5-fluorouracil and adriamycin versus 5-fluorouracil, adriamycin, and polyadenylic-polyuridylic acid (poly A:U) for locally advanced gastric cancer after curative surgery: final results of 15-year follow-up.
Annals of oncology : official journal of the European Society for Medical Oncology (2007-11-22)
H-C Jeung, Y W Moon, S Y Rha, N C Yoo, J K Roh, S H Noh, J S Min, B S Kim, H C Chung
ABSTRACT
This phase III trial was to compare 5-fluorouracil (5-FU), adriamycin, and polyadenylic-polyuridylic acid (poly A:U) against 5-fluorouracil plus adriamycin (FA) for operable gastric cancer. From 1984 to 1989, patients who had D(2-3) curative resection were randomly assigned to receive chemotherapy or chemoimmunotherapy. Chemotherapy consisted of 12 mg/kg 5-FU every week for 18 months and 40 mg/m2 adriamycin every 3 weeks for 12 cycles. Chemoimmunotherapy consisted of FA plus 100 mg of poly A:U weekly for six cycles and was followed 6 months later by six weekly 50-mg booster injections. A total of 292 patients were enrolled. After excluding 12 ineligible patients, 142 and 138 patients were allocated to each treatment. Patients were balanced with prognostic variables: age, sex, tumor location, differentiation, degree of tumor invasion (T2-T4a), and lymph node status (N0-N2). During the 15-year follow-up, chemoimmunotherapy significantly prolonged overall (P = 0.013) and recurrence-free (P = 0.005) survivals compared with chemotherapy alone. The survival benefits were prominent in the subset of patients with T3/T4a, N2, or stage III. Treatments were generally well tolerated in both arms. These results indicate a survival advantage of chemoimmunotherapy with a regimen of FA and poly A:U in curatively resected gastric adenocarcinoma.