- Rotational polymorphism in 2-naphthalenethiol SAMs on Au(111).
Rotational polymorphism in 2-naphthalenethiol SAMs on Au(111).
Journal of the American Chemical Society (2006-09-21)
Peng Jiang, Aymeric Nion, Alexandr Marchenko, Luc Piot, Denis Fichou
PMID16984169
ABSTRACT
We evidence by STM that 2-naphthalenethiol self-assembled monolayers formed at the n-tetradecane/Au(111) interface coexist as two structural phases which both possess molecules into two different orientations (standing and lying). Such a rotational polymorphism is observed and understood at the molecular level for the first time.