Merck
  • Antioxidative activity of sulfur-containing compounds in Allium species for human low-density lipoprotein (LDL) oxidation in vitro.
Antioxidative activity of sulfur-containing compounds in Allium species for human low-density lipoprotein (LDL) oxidation in vitro.
Journal of agricultural and food chemistry (2003-11-13)
Ohki Higuchi, Koutaro Tateshita, Hiroyuki Nishimura
ABSTRACT
Sulfur-containing compounds contributing to health promotion in Allium species are produced via enzymic and thermochemical reactions. Sulfur-containing amino acids and volatile organosulfur compounds were prepared for an antioxidative assay. The inhibitory activity of S-alk(en)yl-l-cysteines and their sulfoxides, volatile alk(en)yl disulfides and trisulfides, and vinyldithiins in Allium species against lipid hydroperoxide (LOOH) formation in human low-density lipoprotein (LDL) was examined. It was elucidated that the alk(en)yl substituents (methyl, propyl, and allyl) and the number of sulfur atoms in the compounds were important for the antioxidative activity. 3,4-Dihydro-3-vinyl-1,2-dithiin, which is produced by a thermochemical reaction of allyl 2-propenethiosulfinate, exhibited the highest antioxidative activity of human LDL among sulfur-containing compounds.