Merck
  • Evaluation of different expression systems for the heterologous expression of pyranose 2-oxidase from Trametes multicolor in E. coli.
Evaluation of different expression systems for the heterologous expression of pyranose 2-oxidase from Trametes multicolor in E. coli.
Microbial cell factories (2010-03-11)
Oliver Spadiut, Gerald Posch, Roland Ludwig, Dietmar Haltrich, Clemens K Peterbauer
ABSTRACT
The heterologous production of the industrially relevant fungal enzyme pyranose 2-oxidase in the prokaryotic host E. coli was investigated using 3 different expression systems, i.e. the well-studied T7 RNA polymerase based pET21d+, the L-arabinose inducible pBAD and the pCOLD system. Preliminary experiments were done in shaking flasks at 25 degrees C and optimized induction conditions to compare the productivity levels of the different expression systems. The pET21d+ and the pCOLD system gave 29 U/L x h and 14 U/L x h of active pyranose 2-oxidase, respectively, whereas the pBAD system only produced 6 U/L x h. Process conditions for batch fermentations were optimized for the pET21d+ and the pCOLD systems in order to reduce the formation of inactive inclusion bodies. The highest productivity rate with the pET21d+ expression system in batch fermentations was determined at 25 degrees C with 32 U/L x h. The pCOLD system showed the highest productivity rate (19 U/L x h) at 25 degrees C and induction from the start of the cultivation. Using the pCOLD system in a fed batch fermentation at 25 degrees C with a specific growth rate of mu = 0.15 h-1resulted in the highest productivity rate of active pyranose oxidase with 206 U/L x h.