Merck
  • Effects of (R)-8-OH-DPAT and the enantiomers of UH-301 on motor activities in the rat: antagonism of (R)-8-OH-DPAT-induced effects.
Effects of (R)-8-OH-DPAT and the enantiomers of UH-301 on motor activities in the rat: antagonism of (R)-8-OH-DPAT-induced effects.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (1992-06-01)
L Björk, A Fredriksson, U Hacksell, T Lewander
ABSTRACT
The effects of the enantiomers of 5-fluoro-8-hydroxy-2-(dipropylamino)tetralin, UH-301 and the potent 5-HT1A-receptor agonist (R)-8-hydroxy-2-(dipropylamino)tetralin, (R)-8-OH-DPAT, on locomotion, rearing and total activity were studied in rats. The experiments were performed as tests either of exploratory activity in non-habituated rats or of motor activity of rats habituated to the environment for 2 h before drug injection. (R)-8-OH-DPAT increased locomotion and total activity and decreased rearing in both conditions. (R)-UH-301 increased locomotion and slightly also total activity in habituated rats and decreased rearing in both conditions. (S)-UH-301 decreased locomotion and rearing in both conditions but only in doses of 10 mumol/kg and above. Lower doses of (S)-UH-301 (10 mumol/kg) antagonized (R)-8-OH-DPAT-induced increases of locomotion and total activity. As (S)-UH-301 decreased rearing, per se, it was not able to antagonize the (R)-8-OH-DPAT induced decrease. These results further support previous data that (S)-UH-301 is a 5-HT1A antagonist while (R)-UH-301 is a 5-HT1A agonist.