Merck
  • A Small-Molecule Oligosaccharyltransferase Inhibitor with Pan-flaviviral Activity.
A Small-Molecule Oligosaccharyltransferase Inhibitor with Pan-flaviviral Activity.
Cell reports (2017-12-16)
Andreas S Puschnik, Caleb D Marceau, Yaw Shin Ooi, Karim Majzoub, Natalie Rinis, Joseph N Contessa, Jan E Carette
ABSTRACT
The mosquito-borne flaviviruses include important human pathogens such as dengue, Zika, West Nile, and yellow fever viruses, which pose a serious threat for global health. Recent genetic screens identified endoplasmic reticulum (ER)-membrane multiprotein complexes, including the oligosaccharyltransferase (OST) complex, as critical flavivirus host factors. Here, we show that a chemical modulator of the OST complex termed NGI-1 has promising antiviral activity against flavivirus infections. We demonstrate that NGI-1 blocks viral RNA replication and that antiviral activity does not depend on inhibition of the N-glycosylation function of the OST. Viral mutants adapted to replicate in cells deficient of the OST complex showed resistance to NGI-1 treatment, reinforcing the on-target activity of NGI-1. Lastly, we show that NGI-1 also has strong antiviral activity in primary and disease-relevant cell types. This study provides an example for advancing from the identification of genetic determinants of infection to a host-directed antiviral compound with broad activity against flaviviruses.
Sigma-Aldrich
Anti-Flavivirus Group Antigen Antibody, clone D1-4G2-4-15, ascites fluid, clone D1-4G2-4-15, Chemicon®