Merck
  • Roles of Signal Transducer Pathways in Investigation of Biopsies from Patients with Bladder Tumors
Roles of Signal Transducer Pathways in Investigation of Biopsies from Patients with Bladder Tumors
Asian Pacific journal of cancer prevention : APJCP (2017-02-28)
Aysegul Bayrak, Sukru Palanduz, Ender Coskunpinar, Oner Sanli, Abdullah Armagan, Serkan Karakus, Ramazan Topaktas, Kivanc Cefle, Sukru Ozturk, Ali Ucur
ABSTRACT
Background:The process of development of bladder cancer features alteration of normal biological conditions caused by changes in molecular pathways. Removing control over regulation of these pathways could lead to changes in signal transduction and abnormal regulation of genes. During tumor formation and progression, genes regulate critical cellular processes, involved in cell cycling, growth and death. Here we evaluated the expression and prognostic importance of FGFR1, HRAS, CCND1, CCND3, STAT3 and FAS genes. Methods: Tumor tissues of 44 patients diagnosed with bladder cancer were investigated for changes in expression levels of FGFR1, HRAS, CCND1, CCND3, FAS and STAT3 genes by the RT-PCR method. Signal transduction pathways and expression of individual genes related to these pathways were analyzed using the “One Sample Test”. Results: There were statistically significant changes in the expression levels of HRAS, CCND1, CCND3 and STAT3, but not FGFR1 and FAS genes. Examination of associations with age, gender, smoking, chemotherapy, tumor grade and tumor growth pattern using the “Independent Samples Test”, showed importance relations between the CCND1 gene and cigarette smoking and sex. Conclusion: Over-expression of HRAS, CCND1, CCND3 and STAT3 genes may play roles in bladder cancer development and progression, while cigarette smoking is significantly associated with CCND1 gene expression and consequently concluded to be contributing to the development of bladder cancer.