Merck
  • In vivo induction of autophagy in splenocytes of C57BL/6 and BALB/c mice infected with ectromelia orthopoxvirus.
In vivo induction of autophagy in splenocytes of C57BL/6 and BALB/c mice infected with ectromelia orthopoxvirus.
Polish journal of veterinary sciences (2013-05-23)
L Martyniszyn, L Szulc-Dabrowska, A Boratyńska-Jasińska, A M Badowska-Kozakiewicz, M G Niemiałtowski
ABSTRACT
Autophagy is a self-degradation process of cellular components. It plays both antiviral and pro-viral roles in the life cycle of different viruses and the pathogenesis of different viral diseases. In this study, we evaluated autophagy induction in splenocytes of ectromelia virus (ECTV)-resistant C57BL/6 and ECTV-susceptible BALB/c mice during infection with the Moscow strain of the ectromelia virus (ECTV-MOS). Autophagy was analyzed using the Western blot method by assessing type II microtubule-associated protein 1 (MAP1) light chain 3 (LC3) and Beclin 1 expression levels relative to beta-actin. Results indicated an increased ratio of LC3-II to beta-actin in splenocytes of C57BL/6 mice only at 7 day post infection (d.p.i.) compared to uninfected animals. LC3-II/beta-actin and Beclin 1/beta-actin ratios in splenocytes of BALB/c mice increased at 5 d.p.i. and remained high until day 14 and 7 p.i., respectively. We confirmed the formation of autophagosome structures in the spleen of BALB/c mice by transmission electron microscopy (TEM). Moreover, autophagy accompanied necrosis in the splenocytes of infected animals. Results suggest that ECTV-MOS induced autophagy, especially in the spleen of the susceptible mouse strain, may support viral replication and promote cell necrosis.