Merck
  • Functional cooperation among Ras, STAT5, and phosphatidylinositol 3-kinase is required for full oncogenic activities of BCR/ABL in K562 cells.
Functional cooperation among Ras, STAT5, and phosphatidylinositol 3-kinase is required for full oncogenic activities of BCR/ABL in K562 cells.
The Journal of biological chemistry (2002-01-10)
Junko Sonoyama, Itaru Matsumura, Sachiko Ezoe, Yusuke Satoh, Xian Zhang, Yoshihisa Kataoka, Emi Takai, Masao Mizuki, Takashi Machii, Hiroshi Wakao, Yuzuru Kanakura
ABSTRACT
BCR/ABL tyrosine kinase generated from the chromosomal translocation t(9;22) causes chronic myelogenous leukemia and acute lymphoblastic leukemia. To examine the roles of BCR/ABL-activated individual signaling molecules and their cooperation in leukemogenesis, we inducibly expressed a dominant negative (DN) form of Ras, phosphatidylinositol 3-kinase, and STAT5 alone or in combination in p210 BCR/ABL-positive K562 cells. The inducibly expressed DN Ras (N17), STAT5 (694F), and DN phosphatidylinositol 3-kinase (Delta p85) inhibited the growth by 90, 55, and 40%, respectively. During the growth inhibition, the expression of cyclin D2 and cyclin D3 was suppressed by N17, 694F, or Delta p85; that of cyclin E by N17; and that of cyclin A by Delta p85. In addition, N17 induced apoptosis in a small proportion of K562, whereas 694F and Delta p85 were hardly effective. In contrast, coexpression of two DN mutants in any combinations induced severe apoptosis. During these cultures, the expression of Bcl-2 was suppressed by N17, 694F, or Delta p85, and that of Bcl-XL by N17. Furthermore, although K562 was resistant to interferon-alpha- and dexamethasone-induced apoptosis, disruption of one pathway by N17, 694F, or Delta p85 sensitized K562 to these reagents. These results suggested that cooperation among these molecules is required for full leukemogenic activities of BCR/ABL.