Merck
  • Deletion of etoposide-induced 2.4 kb transcript (ei24) reduced cell proliferation and aggregate-size in Dictyostelium discoideum.
Deletion of etoposide-induced 2.4 kb transcript (ei24) reduced cell proliferation and aggregate-size in Dictyostelium discoideum.
The International journal of developmental biology (2018-06-08)
Neha Gupta, Shweta Saran
ABSTRACT
The etoposide-induced 2.4 kb transcript (ei24) gene is induced both by p53 and etoposide, an anti-cancer tumour drug. There is no p53 gene present in Dictyostelium discoideum. Thus, the functions of ei24 in the absence of p53 were analysed. Both overexpressor (ei24OE) and knockout (ei24-) mutants were made to study its role during growth, development and differentiation. Additionally, cell cycle and its response to DNA-damage were also analysed. We identified, characterized and elucidated the functions of the ei24 gene in Dictyostelium. In silico analyses demonstrated the conservation across eukaryotes and in situ hybridization showed it to be prestalk-specific. ei24- cells showed reduced cell proliferation and cell-cohesive properties, ultimately forming small-sized aggregates that developed into miniature and stalky fruiting bodies. The ei24OE cells formed fruiting bodies with engorged or double-decker type sori with short stalks. The ei24- cells showed reduced cAMP signalling with lower intracellular cAMP levels resulting in diminished migration of cells along cAMP gradients. Deletion of ei24 resulted in mis-expression of prestalk-specific markers. Cell cycle analysis revealed an increased bias towards the stalk-pathway by ei24- cells and vice-versa for ei24OE cells. EI24 in Dictyostelium functions even in the absence of p53 and is induced in response to both UV-radiation and etoposide treatments. ei24OE cells showed enhanced DNA-damage repair mechanisms. Also, etoposide treatment and overexpression of ei24 caused G2/M arrest in the cell cycle. Our results indicate that EI24 is important for the growth, development and differentiation of Dictyostelium apart from being a DNA-damage response gene.