Merck
  • Detection of novel intracellular alpha-synuclein oligomeric species by fluorescence lifetime imaging.
Detection of novel intracellular alpha-synuclein oligomeric species by fluorescence lifetime imaging.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2006-10-03)
Jochen Klucken, Tiago F Outeiro, Paul Nguyen, Pamela J McLean, Bradley T Hyman
ABSTRACT
Oligomerization and aggregation of alpha-synuclein molecules are believed to play a major role in neuronal dysfunction and loss in Parkinson's disease (PD) and dementia with Lewy bodies. However, alpha-synuclein oligomerization and aggregation have been detected only indirectly in cells using detergent extraction methods. Here, we show for the first time intracellular alpha-synuclein oligomerization using fluorescence lifetime imaging (FLIM). Two forms of alpha-synuclein homomeric interactions were detected: an antiparallel amino terminus-carboxyl terminus interaction between alpha-synuclein molecules, and a close amino terminus-carboxy terminus interaction within single alpha-synuclein molecules. Coexpression of the chaperone protein Hsp70, which can block alpha-synuclein toxicity in several systems, causes alpha-synuclein to adopt a different, open conformation, but Hsp70 does not alter alpha-synuclein-alpha-synuclein interactions. Thus, the neuroprotective effect of Hsp70 can be explained by its chaperone activity on alpha-synuclein molecules, rather than alteration of alpha-synuclein-alpha-synuclein interactions.