Merck
  • Laser irradiation activates spinal adenosine A1 receptor to alleviate osteoarthritis pain in monosodium iodoacetate injected rats.
Laser irradiation activates spinal adenosine A1 receptor to alleviate osteoarthritis pain in monosodium iodoacetate injected rats.
Journal of integrative neuroscience (2020-07-25)
Yuan Li, Fan Wu, Li Xing Lao, Xue Yong Shen
ABSTRACT
The analgesic role of the adenosine A1 receptor is thought to involve the modulation of the spinal N-methyl D-aspartate receptor-mediated nociceptive pathway, which is suggested to be an underlying mechanism in chronic pain. Knee osteoarthritis is a degenerative condition accompanied by chronic pain. We have demonstrated that 10.6-μm laser irradiation has an antinociceptive effect in the monosodium iodoacetate -induced knee osteoarthritis in rats. However, its mechanism of action has yet to be explored. In the present work, we investigate the mechanism of 10.6-μm laser irradiation mediated antinociception in the monosodium iodoacetate -induced knee osteoarthritis. Results showed that the 10.6-μm laser significantly reversed the monosodium iodoacetate -induced nociceptive behaviors for up to 28 days. Moreover, the up-regulation of the A1 receptor and the down-regulated phosphorylation of the N-methyl D-aspartate receptor 1 subunit of the N-methyl D-aspartate receptor were observed in the spinal cord dorsal horn in the monosodium iodoacetate injected rats treated by laser irradiation. Intrathecal injection of 8-cyclopentyl-1,3-dipropylxanthine markedly reversed the effects of laser irradiation, as evidenced both by behavioral pain tests and by levels of spinal phosphorylation of N-methyl D-aspartate receptor 1. These results suggest that the spinal A1 receptor contributes to the antinociceptive effects of 10.6-μm laser, at least in part by inhibiting phosphorylation of N-methyl D-aspartate receptor 1 in the monosodium iodoacetate -induced knee osteoarthritis pain.