Merck
  • CAPN2 acts as an indicator of hepatitis B virus to induce hepatic fibrosis.
CAPN2 acts as an indicator of hepatitis B virus to induce hepatic fibrosis.
Journal of cellular biochemistry (2019-11-05)
Rui Feng, Weixing Du, Ping Lui, Jun Zhang, Yanqing Liu
ABSTRACT
This study is aimed to investigate whether calpain 2 (CAPN2) serves as an indicator of the hepatitis B virus (HBV) to induce hepatic fibrosis. Differentially-expressed genes (DEGs) in HBV-induced hepatic fibrosis and normal liver tissues were analyzed, and signal pathway which was analyzed by the Kyoto Encyclopedia of Genes and Genomes (KEGG) analysis using DEGs. Next, the gene-related network map was constructed using the Search Tool for the Retrieval of Interacting Genes. Moreover, CAPN2 protein expression, level of hepatic fibrosis, CAPN2 messenger RNA level, and protein levels of CAPN2, a-SAM, COL3A1, COL1A1, and MAPK1 were determined using Immunohistochemistry (IHC), hematoxylin and eosin, RT-qPCR, and western blot (WB), respectively. There were 420 DEGs screened in HBV-induced hepatic fibrosis and normal liver tissues, among which, 373 were significantly upregulated and 47 were obviously downregulated. KEGG analysis showed that the upregulated DEGs were mainly concentrated in extracellular matrix-receptor interaction, protein digestion, and absorption signaling pathways. The network diagram analysis showed that the DEGs, such as CAPN2, ITGAV, and CCR2, play the key role in the DEG network map, and CAPN2 related to hepatic fibrosis via MAPK1. The increased CAPN2 expression and obvious hepatic fibrosis was displayed in the HBV-induced hepatic fibrosis tissues. In addition, HBV could induce CAPN2 expression, and the interference of CAPN2 could inhibit the expression of hepatic fibrosis markers, including a-SAM, COL3A1, COL1A1, and MAPK1. CAPN2 is regarded as a biomarker of hepatic fibrosis induced by HBV.