Merck
  • Cytomegalovirus infection and associated hospitalization and costs among individuals undergoing allogeneic hematopoietic stem cell transplant.
Cytomegalovirus infection and associated hospitalization and costs among individuals undergoing allogeneic hematopoietic stem cell transplant.
Current medical research and opinion (2019-09-07)
Jonathan Schelfhout, Harold Brown, John A House, Amit D Raval
ABSTRACT
Objective: This study utilized a large, national US database to explore the impact of Cytomegalovirus (CMV) infection on hospital services utilization and costs during the first 100 days following allogeneic hematopoietic stem cell transplant (allo-HSCT).Methods: This retrospective, observational cohort study used data from the Premier Healthcare database to identify patients undergoing their first (index) allo-HSCT procedure between 1 January 2006 and 31 March 2015. Three subgroups were analyzed according to CMV-related readmissions during the 100-day follow-up (0, 1, or 2+ readmissions) to compare healthcare utilization and costs.Results: A total of 1610 patients (mean age, 50.5 years; 56.9% male) from 52 US hospitals met the inclusion criteria. During follow-up, 212 (13.2%) patients had 1 (n = 161; 10.0%) or 2+ (n = 51; 3.2%) CMV-related readmissions. The mean ± SD number of all follow-up encounters (inpatient admissions and hospital-based outpatient visits) was similar for the no CMV (3.9 ± 3.9), 1 CMV (3.7 ± 3.9), and 2+ CMV (4.5 ± 3.8) readmission groups (p = .439). Mean total costs of hospital-based healthcare encounters (inpatient admissions and hospital-based outpatient visits) during follow-up were significantly greater in patients who had a CMV readmission ($111,729 [1 CMV readmission]; $184,021 [2+ CMV readmissions]) compared to those without a CMV readmission ($46,064; p < .001]. 100-day follow-up mortality was higher in the CMV vs. non-CMV readmission groups (30.2 vs. 8.2%; p < .001).Conclusions: This large, national database study revealed significantly higher healthcare utilization and costs, as well as mortality, among patients with CMV-related re-hospitalization during the first 100 days post-transplant as compared to patients without CMV-related hospitalization.