Merck
  • Precision-Cut Tissue Slices (PCTS) from the digestive gland of the Mediterranean mussel Mytilus galloprovincialis: An ex vivo approach for molecular and cellular responses in marine invertebrates.
Precision-Cut Tissue Slices (PCTS) from the digestive gland of the Mediterranean mussel Mytilus galloprovincialis: An ex vivo approach for molecular and cellular responses in marine invertebrates.
Toxicology in vitro : an international journal published in association with BIBRA (2019-07-23)
Maria Elisa Giuliani, Erica Sparaventi, Ilaria Lanzoni, Lucia Pittura, Francesco Regoli, Stefania Gorbi
ABSTRACT
The precision-cut tissue slices (PCTS) represent a largely used biological model in mammalian research. This ex vivo approach offers the main advantages of in vitro systems, while maintaining the natural architecture of the tissue. The use of PCTS in toxicological research has been proposed for investigating the cellular effects of xenobiotics or bioactive compounds mostly in mammalian models. Their application is increasing also in marine organisms, but still limited to fish. This work validates the use of PCTS in an invertebrate species, the Mediterranean mussel Mytilus galloprovincialis. Intact tissue slices of different thicknesses (300, 350 and 400 μm) were successfully obtained from the digestive gland. The slices maintained the histological integrity and the viability after 6 h and 24 h incubation in culture medium, with some differences depending on the thickness. The enzymatic activities and mRNA levels of catalase and glutathione S-transferase, chosen as model biological endpoints, were measured until 24 h incubation, revealing the functionality of such systems. This work demonstrates the suitability of mussel PCTS for investigating molecular and cellular responses in ecotoxicological research.