Merck
  • Controlling micropollutants in tertiary municipal wastewater by O3/H2O2, granular biofiltration and UV254/H2O2 for potable reuse applications.
Controlling micropollutants in tertiary municipal wastewater by O3/H2O2, granular biofiltration and UV254/H2O2 for potable reuse applications.
Chemosphere (2019-09-13)
F Piras, O Santoro, T Pastore, I Pio, E De Dominicis, E Gritti, R Caricato, M G Lionetto, G Mele, D Santoro
ABSTRACT
A comprehensive pilot study was carried out to experimentally assess the potential of newly developed treatment trains integrating two-stage AOPs and biofiltration to reach potable reuse water quality standards from municipal wastewater. The processes consisted of a two-stage AOPs with (carbon or limestone) biofiltration, the first AOP (O3/H2O2) serving as pre-treatment to biofiltration and the second AOP (UV254/H2O2) serving as post-biofiltration finishing step to ensure advanced disinfection. A comprehensive monitoring campaign was put in place resulting from the combination of targeted, non-targeted and suspect screening measurements. It was found that 13 organic micropollutants were detected from a list of 219 suspects although at ng/L level only. For the treatment conditions piloted in this study (O3 = 13 ± 0.5 mg/L, H2O2 = 11 ± 0.4 mg/L for the O3/H2O2 process, and UV = 410 ± 63.5 mJ/cm2, H2O2 = 5 mg/l for the UV254/H2O2 process), it was possible to estimate the overall removal efficacy for each unit process, which was found to follow this order: RO (99%)  > BAC (87%) > O3-H2O2 (78%)  > BAL (67%)  > UV/H2O2 (43%)  > AOP contact chamber (19%)  > UF(0%), with the treatment train integrating two AOPs and granular biofiltration with activated carbon (O3/H2O2 + BAC + UV254/H2O2) showing superior performance with a 99% abatement in total micropollutants. No ecotoxicologically-positive response was generally observed for any of the effluent samples from the tested trains, even when pre-concentration factors up to 100-1000 times were employed to increase the sensitivity of the bioassay methods.