Merck
  • Down-regulation of dopamine D-2, 5-HT2 receptors and beta-adrenoceptors in rat brain after prolonged treatment with a new potential antidepressant, Lu 19-005.
Down-regulation of dopamine D-2, 5-HT2 receptors and beta-adrenoceptors in rat brain after prolonged treatment with a new potential antidepressant, Lu 19-005.
Journal of neural transmission (1985-01-01)
G Nowak, J Arnt, J Hyttel, O Svendsen
ABSTRACT
Lu 19-005 is a new phenylindan derivative with strong and equipotent inhibitory effect on dopamine (DA), noradrenaline (NA) and serotonin (5-HT) uptake. The adaptive effects of 2 weeks treatment with Lu 19-005, on receptor binding in vitro and on d-amphetamine responsiveness in vivo have been investigated in rats. One or 3 days after the final dose the number of beta-adrenoceptors and of 5-HT2 and DA D-2 receptors was decreased by 20-30%, whereas alpha 1-adrenoceptor number was slightly decreased only 1 day after withdrawal. The DA D-2 receptor number remained decreased at 7 days withdrawal, but returned to normal after another 3 days. The brain levels of DA, NA and 5-HT were not changed by 2 weeks' Lu 19-005 treatment. The down-regulation of DA D-2 receptors was accompanied by tolerance to d-amphetamine-induced hypermotility (after low doses) and stereotyped licking or biting (after a high dose). The tolerance to d-amphetamine-induced hypermotility was maximal at 3-5 days withdrawal time, and remained significant also 15 days after the last dose. An acute dose of Lu 19-005 did not modify the effects of d-amphetamine. The results are discussed in relation to the effect of prolonged treatment with other antidepressant drugs.