Merck
  • Cloning and characterization of hELD/OSA1, a novel BRG1 interacting protein.
Cloning and characterization of hELD/OSA1, a novel BRG1 interacting protein.
The Biochemical journal (2002-05-04)
Adam F L Hurlstone, Ivan A Olave, Nick Barker, Mascha van Noort, Hans Clevers
ABSTRACT
A highly conserved multisubunit enzymic complex, SWI/SNF, participates in the regulation of eukaryote gene expression through its ability to remodel chromatin. While a single component of SWI/SNF, Swi2 or a related protein, can perform this function in vitro, the other components appear to modulate the activity and specificity of the complex in vivo. Here we describe the cloning of hELD/OSA1, a 189 KDa human homologue of Drosophila Eld/Osa protein, a constituent of Drosophila SWI/SNF. By comparing conserved peptide sequences in Eld/Osa homologues we define three domains common to all family members. A putative DNA binding domain, or ARID (AT-rich DNA-interacting domain), may function in targetting SWI/SNF to chromatin. Two other domains unique to Eld/Osa proteins, EHD1 and EHD2, map to the C-terminus. We show that EHD2 mediates binding to Brahma-related gene 1 (BRG1), a human homologue of yeast Swi2. EHD1 and EHD2 also appear capable of interacting with each other. Using an antibody raised against EHD2 of hELD/OSA1, we detected Eld/Osa1 in endogenous SWI/SNF complexes derived from mouse brain.