Merck
Aldosterone inhibits antifibrotic factors in mouse hypertensive heart.
Hypertension (Dallas, Tex. : 1979) (2012-05-02)
Feriel Azibani, Ludovic Benard, Saskia Schlossarek, Regine Merval, François Tournoux, Loubina Fazal, Evelyne Polidano, Jean-Marie Launay, Lucie Carrier, Christos Chatziantoniou, Jane-Lise Samuel, Claude Delcayre
ABSTRACT
The renin-angiotensin-aldosterone system is involved in the arterial hypertension-associated cardiovascular remodeling. In this context, the development of cardiac fibrosis results from an imbalance between profibrotic and antifibrotic pathways, in which the role of aldosterone is yet not established. To determine the role of intracardiac aldosterone in the development of myocardial fibrosis during hypertension, we used a double transgenic model (AS-Ren) of cardiac hyperaldosteronism (AS) and systemic hypertension (Ren). The 9-month-old hypertensive mice had cardiac fibrosis, and hyperaldosteronism enhanced the fibrotic level. The mRNA levels of connective tissue growth factor and transforming growth factor-β1 were similarly increased in Ren and AS-Ren mice compared with wild-type and AS mice, respectively. Hyperaldosteronism combined with hypertension favored the macrophage infiltration (CD68(+) cells) in heart, and enhanced the mRNA level of monocyte chemoattractant protein 1, osteopontin, and galectin 3. Interestingly, in AS-Ren mice the hypertension-induced increase in bone morphogenetic protein 4 mRNA and protein levels was significantly inhibited, and B-type natriuretic peptide expression was blunted. The mineralocorticoid receptor antagonist eplerenone restored B-type natriuretic peptide and bone morphogenetic protein 4 levels and decreased CD68 and galectin 3 levels in AS-Ren mice. Finally, when hypertension was induced by angiotensin II infusion in wild-type and AS mice, the mRNA profiles did not differ from those observed in Ren and AS-Ren mice, respectively. The aldosterone-induced inhibition of B-type natriuretic peptide and bone morphogenetic protein 4 expression was confirmed in vitro in neonatal mouse cardiomyocytes. Altogether, we demonstrate that, at the cardiac level, hyperaldosteronism worsens hypertension-induced fibrosis through 2 mineralocorticoid receptor-dependent mechanisms, activation of inflammation/galectin 3-induced fibrosis and inhibition of antifibrotic factors (B-type natriuretic peptide and bone morphogenetic protein 4).