Merck
  • Asymmetric Rh(I)-catalyzed addition of MIDA boronates to N-tert-butanesulfinyl aldimines: development and comparison to trifluoroborates.
Asymmetric Rh(I)-catalyzed addition of MIDA boronates to N-tert-butanesulfinyl aldimines: development and comparison to trifluoroborates.
The Journal of organic chemistry (2010-04-15)
Katrien Brak, Jonathan A Ellman
ABSTRACT
The Rh(I)-catalyzed addition of alkenyl and aryl MIDA boronates to N-tert-butanesulfinyl aromatic and aliphatic imines proceeds in good yields (up to 99%) and with very high selectivity (98:2 to >99:1). In comparison to trifluoroborates, higher yields and selectivities are observed for the addition to N-tert-butanesulfinyl aromatic imines. This new method expands upon the versatility of the Rh(I)-catalyzed addition of boron reagents to imines, thereby further enabling the synthesis of chiral alpha-branched amines.