Merck
  • Exposure to Zinc oxide nanoparticles during pregnancy induces oocyte DNA damage and affects ovarian reserve of mouse offspring.
Exposure to Zinc oxide nanoparticles during pregnancy induces oocyte DNA damage and affects ovarian reserve of mouse offspring.
Aging (2018-08-29)
Qiu-Yue Zhai, Wei Ge, Jun-Jie Wang, Xiao-Feng Sun, Jin-Mei Ma, Jing-Cai Liu, Yong Zhao, Yan-Zhong Feng, Paul W Dyce, Massimo De Felici, Wei Shen
ABSTRACT
Zinc oxide nanoparticles (nZnO) have been shown to have higher toxic effects likely due to their ion-shedding ability and low solubility under neutral conditions. In order to investigate whether exposure to nZnO during embryonic development affects ovary development, 12.5 day post coitum (dpc) fetal mouse ovaries were cultured in the presence of nZnO for 6 days. We found that the nanoparticles (NPs) accumulated within the oocyte cytoplasm in a dose dependent manner, caused DNA damage and apoptosis, and result in a significant decrease in oocyte numbers. No such effects were observed when the ovaries were incubated in the presence of ZnSO4 or bulk ZnO as controls. In addition, we injected intravenously 16 mg/kg body weight nZnO in 12.5 dpc pregnant mice on two consecutive days and analyzed the ovaries of fetuses or offspring at three critical periods of oogenesis: 17.5 dpc, 3 days post-partum (dpp) and 21 dpp. Evidence of increased DNA damage in pachytene oocytes in fetal ovaries and impaired primordial follicle assembly and folliculogenesis dynamics in the ovaries of the offspring were found. Our results indicate that certain types of NPs affect pre- and post-natal oogenesis in vitro and in vivo.