Merck
  • Comparative Early Life Stage Toxicity of the African Clawed Frog, Xenopus laevis Following Exposure to Selected Herbicide Formulations Applied to Eradicate Alien Plants in South Africa.
Comparative Early Life Stage Toxicity of the African Clawed Frog, Xenopus laevis Following Exposure to Selected Herbicide Formulations Applied to Eradicate Alien Plants in South Africa.
Archives of environmental contamination and toxicology (2017-10-21)
Oluwaseun O Babalola, J H Van Wyk
ABSTRACT
The rise in pesticides application has increased the need for better understanding of their ecological impacts. The global amphibian declines, for example, have been positively correlated with pesticides use. The differential susceptibility in the developmental stages of amphibians to chemical substances are still largely unknown. We examined the 96-h differential toxicity responses of embryos, premetamorphic and transitional larval stage of Xenopus laevis, to six formulated aquatic herbicide products containing the active ingredients of diquat dibromide (Midstream), glufosinate ammonium (Basta), imazapyr (Arsenal), and three glyphosate formulations (Roundup, Kilo Max, and Environ Glyphosate). The results showed the premetamorphic stage as the most sensitive to the herbicides toxicity. This study confirmed that the developmental stage at which amphibian are exposed to contaminants is critical to their survival and that the chemical contamination hypothesis of the global decline of amphibians should continue to be considered. This establishment of the premetamorphic larval as sensitive toxicity representative for all developmental stages of X. laevis means that this stage could be used more extensively in pesticides toxicity assessments.