Skip to Content
Merck
L-Ascorbic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ascorbic Acid

L-Ascorbic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
L-Ascorbic acid, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique