Skip to Content
Merck
(−)-Nicotine PESTANAL®, analytical standard

(−)-Nicotine

(−)-Nicotine PESTANAL®, analytical standard
Synonym(s):
(−)-1-Methyl-2-(3-pyridyl)pyrrolidine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing