
Residual Solvent - Trichloroethylene

Synonym(s):
Trichloroethylene, TCE, Trichloroethene
Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Products
Brand
Boiling Point (°C)
Formula Weight
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing