Skip to Content
Merck
Hydrogen peroxide solution 31% Ultrapur

Hydrogen peroxide solution 31%

Hydrogen peroxide solution 31% Ultrapur
Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing