
Everolimus for system suitability

Synonym(s):
Everolimus, 42-O-(2-Hydroxyethyl)rapamycin
Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing