Skip to Content
Merck
Tetrachloroauric(III) acid trihydrate 99% for analysis EMSURE®

Gold(III) chloride trihydrate

Tetrachloroauric(III) acid trihydrate 99% for analysis EMSURE®
Synonym(s):
Hydrogen tetrachloroaurate(III), Tetrachloroauric(III) acid, Tetrachloroauric(III) acid trihydrate 99%
Linear Formula:
HAuCl4 · 3H2O
CAS Number:
Molecular Weight:
393.83

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Package Size
Reagent Type
Core
Special Grade