Cetylpyridinium chloride
Synonym(s):
Hexadecylpyridinium chloride monohydrate, Cetylpyridinium chloride monohydrate, 1-Cetylpyridinium chloride monohydrate
Empirical Formula (Hill Notation):
C21H38ClN · xH2O
CAS Number:
Molecular Weight:
339.99 (anhydrous basis)
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing