Skip to Content
Merck
2,6-Dimethylbenzenethiol ≥95%, FG

2,6-Dimethylbenzenethiol

2,6-Dimethylbenzenethiol ≥95%, FG
Synonym(s):
2,6-Dimethylthiophenol
Linear Formula:
(CH3)2C6H3SH
CAS Number:
Molecular Weight:
138.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing