Skip to Content
Merck
Copper(II) sulfate ReagentPlus®, ≥99%

Copper(II) sulfate

Copper(II) sulfate ReagentPlus®, ≥99%
Synonym(s):
Cupric sulfate, Copper(II) sulfate anhydrous
Linear Formula:
CuSO4
CAS Number:
Molecular Weight:
159.61

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing