Skip to Content
PH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Drierite
™
Synonym(s):
Calcium sulfate, Gypsum
Linear Formula:
CaSO
4
CAS Number:
7778-18-9
Molecular Weight:
136.14
EC Number:
231-900-3
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
238988
Drierite
™
,
with indicator, 8 mesh
Expand
View Pricing
238961
Drierite
™
,
with indicator, 4 mesh
Expand
View Pricing
238910
Drierite
™
,
without indicator, 4 mesh
Expand
View Pricing