Sodium pyruvate
Synonym(s):
α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt
Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Products
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-3 of 3